Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SIN3B
Entrez Gene ID
23309
Gene Name
SIN3 transcription regulator family member B
Gene Aliases
-
Chromosome Location
Chr.19: 16829387 - 16880355 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297595.1 NP_001284524.1 8 - 9 1094 84
NM_015260.3 NP_056075.1 8 - 9 1094 84
GenBank mRNA AB014600.2 - 8 - 9 1081 84
AK001633.1 - 2 - 3 195 84
AY706204.1 - 8 - 9 1074 84

View Full Product Details