Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DIP2A
Entrez Gene ID
23181
Gene Name
disco interacting protein 2 homolog A
Gene Aliases
C21orf106, DIP2
Chromosome Location
Chr.21: 46458942 - 46570900 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146115.1 NP_001139587.1 10 - 11 1483 111
NM_001146116.1 NP_001139588.1 11 - 12 1600 111
NM_015151.3 NP_055966.2 11 - 12 1612 111
NM_206889.2 NP_996772.1 11 - 12 1612 111
NM_206890.2 NP_996773.1 11 - 12 1612 111
NM_206891.2 NP_996774.1 11 - 12 1612 111
XM_011529492.2 XP_011527794.1 12 - 13 1727 111
XM_011529501.1 XP_011527803.1 12 - 13 1727 111
XM_011529502.1 XP_011527804.1 12 - 13 1727 111
XM_017028291.1 XP_016883780.1 12 - 13 1727 111
XM_017028292.1 XP_016883781.1 12 - 13 1727 111
XM_017028293.1 XP_016883782.1 12 - 13 1724 111
XM_017028294.1 XP_016883783.1 11 - 12 1606 111
XM_017028295.1 XP_016883784.1 11 - 12 1603 111
XM_017028296.1 XP_016883785.1 11 - 12 1603 111
XM_017028297.1 XP_016883786.1 11 - 12 1595 111
XM_017028298.1 XP_016883787.1 10 - 11 1474 111
XM_017028299.1 XP_016883788.1 11 - 12 1472 111
XM_017028300.1 XP_016883789.1 12 - 13 1727 111
XM_017028301.1 XP_016883790.1 12 - 13 1727 111
XM_017028302.1 XP_016883791.1 10 - 11 1367 111
XM_017028303.1 XP_016883792.1 12 - 13 1727 111
XM_017028304.1 XP_016883793.1 12 - 13 1727 111
XM_017028305.1 XP_016883794.1 12 - 13 1727 111
GenBank mRNA AB273729.1 - 11 - 12 1450 111
AF432263.1 - 6 - 7 822 111
AF490768.1 - 11 - 12 1505 111
AK056738.1 - 11 - 12 1504 111
AK098236.1 - 6 - 7 755 111
AK303351.1 - 9 - 10 1427 111
BC033718.1 - 10 - 11 1466 111
BC038443.1 - 11 - 12 1611 111

View Full Product Details