Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CAPRIN2
Entrez Gene ID
65981
Gene Name
caprin family member 2
Gene Aliases
C1QDC1, EEG-1, EEG1, RNG140
Chromosome Location
Chr.12: 30709552 - 30754994 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002259.2 NP_001002259.1 5 - 6 1642 112
NM_001206856.2 NP_001193785.1 5 - 6 1642 112
NM_001319843.1 NP_001306772.1 5 - 6 1642 112
NM_001319844.1 NP_001306773.1 6 - 7 897 112
NM_001319845.1 NP_001306774.1 6 - 7 897 112
NM_001319846.1 NP_001306775.1 6 - 7 887 112
NM_023925.4 NP_076414.2 5 - 6 1642 112
NM_032156.4 NP_115532.3 5 - 6 1642 112
XM_006719138.3 XP_006719201.1 5 - 6 1765 112
XM_006719139.3 XP_006719202.1 5 - 6 1765 112
XM_006719140.3 XP_006719203.1 5 - 6 1765 112
XM_006719142.3 XP_006719205.1 5 - 6 1765 112
XM_006719144.3 XP_006719207.1 5 - 6 1765 112
XM_006719145.3 XP_006719208.1 5 - 6 1765 112
XM_006719146.3 XP_006719209.1 5 - 6 1765 112
XM_006719147.3 XP_006719210.1 5 - 6 1765 112
XM_017019851.1 XP_016875340.1 5 - 6 1765 112
XM_017019852.1 XP_016875341.1 6 - 7 860 112
XM_017019853.1 XP_016875342.1 6 - 7 930 112
XM_017019854.1 XP_016875343.1 5 - 6 1765 112
XM_017019855.1 XP_016875344.1 6 - 7 887 112
XM_017019856.1 XP_016875345.1 6 - 7 887 112
XM_017019857.1 XP_016875346.1 6 - 7 860 112
XM_017019858.1 XP_016875347.1 6 - 7 887 112
XM_017019859.1 XP_016875348.1 6 - 7 860 112
XM_017019860.1 XP_016875349.1 5 - 6 1765 112
XM_017019861.1 XP_016875350.1 6 - 7 887 112
XM_017019862.1 XP_016875351.1 5 - 6 1765 112
XM_017019863.1 XP_016875352.1 5 - 6 1765 112
XM_017019864.1 XP_016875353.1 5 - 6 1765 112
XM_017019865.1 XP_016875354.1 5 - 6 1765 112
XM_017019866.1 XP_016875355.1 6 - 7 860 112
XM_017019867.1 XP_016875356.1 6 - 7 887 112
XM_017019868.1 XP_016875357.1 6 - 7 860 112
XM_017019869.1 XP_016875358.1 6 - 7 887 112
XM_017019870.1 XP_016875359.1 6 - 7 860 112
XM_017019871.1 XP_016875360.1 6 - 7 931 112
XM_017019872.1 XP_016875361.1 6 - 7 887 112
XM_017019873.1 XP_016875362.1 6 - 7 887 112
XM_017019875.1 XP_016875364.1 6 - 7 887 112
GenBank mRNA AL834212.1 - 2 - 3 129 112
AY074490.1 - 5 - 6 1642 112
BC117672.1 - 5 - 6 1439 112
BC117673.1 - 5 - 6 1439 112

View Full Product Details