The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256041.1 | NP_001242970.1 | 22 - 23 | 2564 | 73 |
NM_005379.3 | NP_005370.1 | 21 - 22 | 2527 | 73 | |
XM_011538373.2 | XP_011536675.1 | 21 - 22 | 2495 | 73 | |
GenBank mRNA | AF009961.1 | - | 20 - 21 | 2276 | 73 |
AF105424.1 | - | 21 - 22 | 2485 | 73 | |
AF127026.1 | - | 21 - 22 | 2520 | 73 | |
AK300381.1 | - | 17 - 18 | 73 | ||
AK312431.1 | - | 22 - 23 | 2564 | 73 | |
BC059387.1 | - | 21 - 22 | 2527 | 73 |