Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEMA3D
Entrez Gene ID
223117
Gene Name
semaphorin 3D
Gene Aliases
Sema-Z2, coll-2
Chromosome Location
Chr.7: 84995192 - 85187176 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152754.2 NP_689967.2 15 - 16 1804 143
XM_011515960.1 XP_011514262.1 17 - 18 2114 143
XM_011515961.2 XP_011514263.1 15 - 16 1639 143
XM_017011873.1 XP_016867362.1 16 - 17 2303 143
GenBank mRNA AY358937.1 - 16 - 17 1870 143
BC029590.1 - 8 - 9 890 143

View Full Product Details