The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145159.2 | NP_660142.1 | 9 - 10 | 1673 | 115 |
GenBank mRNA | AF020201.1 | - | 9 - 10 | 1320 | 229 |
AF029779.1 | - | 9 - 10 | 1673 | 115 |