Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SGIP1
Entrez Gene ID
84251
Gene Name
SH3 domain GRB2 like endophilin interacting protein 1
Gene Aliases
-
Chromosome Location
Chr.1: 66533272 - 66751139 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308203.1 NP_001295132.1 15 - 16 1259 69
XM_006710961.2 XP_006711024.1 18 - 19 1876 69
XM_006710971.2 XP_006711034.1 17 - 18 1375 69
XM_006710973.2 XP_006711036.1 15 - 16 1351 69
XM_006710974.2 XP_006711037.1 14 - 15 1267 69
XM_017002525.1 XP_016858014.1 16 - 17 1454 69
XM_017002532.1 XP_016858021.1 15 - 16 1279 69
GenBank mRNA AK090697.1 - 9 - 10 816 69
BX640813.1 - 15 - 16 1236 69

View Full Product Details