The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019849.2 | NP_062823.1 | 1 - 2 | 297 | 64 |
XM_006723284.2 | XP_006723347.1 | 1 - 2 | 169 | 64 | |
XM_011527119.1 | XP_011525421.1 | 1 - 2 | 169 | 64 | |
GenBank mRNA | AB037670.1 | - | 1 - 2 | 249 | 64 |
AJ277731.1 | - | 1 - 2 | 258 | 64 | |
AK316594.1 | - | 1 - 2 | 169 | 64 | |
BC035627.1 | - | 1 - 2 | 240 | 64 |