Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC82
Entrez Gene ID
79780
Gene Name
coiled-coil domain containing 82
Gene Aliases
HSPC048
Chromosome Location
Chr.11: 96352765 - 96389940 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318736.1 NP_001305665.1 3 - 4 289 74
NM_001318737.1 NP_001305666.1 2 - 3 154 74
NM_024725.3 NP_079001.2 2 - 3 154 74
XM_006718911.3 XP_006718974.1 2 - 3 175 74
XM_011542984.2 XP_011541286.1 2 - 3 818 74
XM_011542987.2 XP_011541289.1 2 - 3 166 74
XM_017018305.1 XP_016873794.1 1 - 2 1899 74
XM_017018306.1 XP_016873795.1 1 - 2 1899 74
XM_017018307.1 XP_016873796.1 2 - 3 541 74
XM_017018311.1 XP_016873800.1 1 - 2 1899 74
XM_017018312.1 XP_016873801.1 2 - 3 818 74
GenBank mRNA AF245436.1 - 2 - 3 142 74
AK056790.1 - 2 - 3 147 74
AK074306.1 - 3 - 4 283 74
BC018663.2 - 1 - 2 89 74
BP219605.1 - 2 - 3 179 74

View Full Product Details