The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020474.3 | NP_065207.2 | 5 - 6 | 956 | 121 |
XM_005258239.3 | XP_005258296.1 | 6 - 7 | 1024 | 121 | |
XM_017025692.1 | XP_016881181.1 | 6 - 7 | 1203 | 121 | |
GenBank mRNA | AK293062.1 | - | 7 - 8 | 1098 | 121 |
BC038440.1 | - | 6 - 7 | 1021 | 121 | |
U41514.1 | - | 5 - 6 | 893 | 121 | |
X85018.1 | - | 5 - 6 | 862 | 121 |