Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPM1D
Entrez Gene ID
8493
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1D
Gene Aliases
PP2C-DELTA, WIP1
Chromosome Location
Chr.17: 60600183 - 60666280 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003620.3 NP_003611.1 5 - 6 1497 68
GenBank mRNA AK312902.1 - 5 - 6 1487 68
BC016480.1 - 5 - 6 1413 68
BC033893.2 - 5 - 6 1371 68
BC060877.1 - 5 - 6 1497 68
BT009780.1 - 5 - 6 1265 68
KU178420.1 - 5 - 6 1265 68
KU178421.1 - 4 - 5 1036 68
U78305.1 - 5 - 6 1468 68

View Full Product Details