Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GLS
Entrez Gene ID
2744
Gene Name
glutaminase
Gene Aliases
AAD20, GAC, GAM, GLS1, KGA
Chromosome Location
Chr.2: 190880821 - 190965552 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256310.1 NP_001243239.1 8 - 9 1341 130
NM_014905.4 NP_055720.3 8 - 9 1341 130
XM_005246467.3 XP_005246524.1 8 - 9 1341 130
XM_006712435.1 XP_006712498.1 8 - 9 1341 130
GenBank mRNA AB020645.1 - 8 - 9 1327 130
AF097492.1 - 8 - 9 975 130
AF158555.1 - 9 - 10 1321 130
AF223943.1 - 8 - 9 1096 130
AF279697.1 - 5 - 6 426 130
AF327434.1 - 8 - 9 1307 130
AK001220.1 - 5 - 6 413 130
AK289747.1 - 9 - 10 1317 130
BC038507.1 - 9 - 10 1317 130

View Full Product Details