The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001976.2 | NP_001001976.1 | 10 - 11 | 1370 | 125 |
NM_001288734.1 | NP_001275663.1 | 9 - 10 | 1266 | 125 | |
NM_001288735.1 | NP_001275664.1 | 8 - 9 | 1243 | 125 | |
NM_001288736.1 | NP_001275665.1 | 10 - 11 | 1605 | 125 | |
NM_007041.3 | NP_008972.2 | 10 - 11 | 1370 | 125 | |
NR_110006.1 | 8 - 9 | 1312 | 125 | ||
XM_005269458.2 | XP_005269515.1 | 11 - 12 | 1511 | 125 | |
XM_005269459.3 | XP_005269516.1 | 11 - 12 | 1734 | 125 | |
XM_017015597.1 | XP_016871086.1 | 8 - 9 | 1180 | 125 | |
XM_017015598.1 | XP_016871087.1 | 10 - 11 | 1589 | 125 | |
XM_017015599.1 | XP_016871088.1 | 9 - 10 | 1253 | 125 | |
XM_017015600.1 | XP_016871089.1 | 10 - 11 | 1233 | 125 | |
XM_017015602.1 | XP_016871091.1 | 9 - 10 | 1150 | 125 | |
XM_017015604.1 | XP_016871093.1 | 8 - 9 | 1095 | 125 | |
GenBank mRNA | AF079098.1 | - | 10 - 11 | 1244 | 125 |
AF079099.1 | - | 10 - 11 | 1244 | 125 | |
AK124669.1 | - | 8 - 9 | 1320 | 125 | |
AK295642.1 | - | 5 - 6 | 602 | 125 | |
AK296356.1 | - | 9 - 10 | 1104 | 125 | |
AK299101.1 | - | 10 - 11 | 1256 | 125 | |
AK303604.1 | - | 8 - 9 | 1201 | 125 | |
AK308926.1 | - | 3 - 4 | 399 | 125 | |
AK313057.1 | - | 10 - 11 | 1330 | 125 | |
BC022026.1 | - | 10 - 11 | 1343 | 125 |