The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001976.2 | NP_001001976.1 | 11 - 12 | 1495 | 88 |
NM_001288734.1 | NP_001275663.1 | 10 - 11 | 1391 | 88 | |
NM_001288735.1 | NP_001275664.1 | 9 - 10 | 1368 | 88 | |
NM_001288736.1 | NP_001275665.1 | 11 - 12 | 1730 | 88 | |
NM_007041.3 | NP_008972.2 | 11 - 12 | 1495 | 88 | |
NR_110006.1 | 9 - 10 | 1437 | 88 | ||
XM_005269458.2 | XP_005269515.1 | 12 - 13 | 1636 | 88 | |
XM_005269459.3 | XP_005269516.1 | 12 - 13 | 1859 | 88 | |
XM_017015597.1 | XP_016871086.1 | 9 - 10 | 1305 | 88 | |
XM_017015598.1 | XP_016871087.1 | 11 - 12 | 1714 | 88 | |
XM_017015599.1 | XP_016871088.1 | 10 - 11 | 1378 | 88 | |
XM_017015600.1 | XP_016871089.1 | 11 - 12 | 1358 | 88 | |
XM_017015602.1 | XP_016871091.1 | 10 - 11 | 1275 | 88 | |
XM_017015604.1 | XP_016871093.1 | 9 - 10 | 1220 | 88 | |
GenBank mRNA | AF079098.1 | - | 11 - 12 | 1369 | 88 |
AF079099.1 | - | 11 - 12 | 1369 | 88 | |
AK292657.1 | - | 6 - 7 | 88 | ||
AK295642.1 | - | 6 - 7 | 88 | ||
AK296356.1 | - | 10 - 11 | 88 | ||
AK299101.1 | - | 11 - 12 | 1381 | 88 | |
AK303604.1 | - | 9 - 10 | 88 | ||
AK308926.1 | - | 4 - 5 | 524 | 88 | |
AK313057.1 | - | 11 - 12 | 1455 | 88 | |
BC022026.1 | - | 11 - 12 | 1468 | 88 |