The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170460.1 | NP_001163931.1 | 15 - 16 | 1757 | 81 |
NM_006201.4 | NP_006192.1 | 15 - 16 | 1892 | 81 | |
NM_033018.3 | NP_148978.2 | 15 - 16 | 1999 | 81 | |
XM_017029569.1 | XP_016885058.1 | 16 - 17 | 2164 | 81 | |
XM_017029570.1 | XP_016885059.1 | 16 - 17 | 1974 | 81 | |
XM_017029571.1 | XP_016885060.1 | 15 - 16 | 2935 | 81 | |
XM_017029572.1 | XP_016885061.1 | 16 - 17 | 2012 | 81 | |
XM_017029573.1 | XP_016885062.1 | 16 - 17 | 2367 | 81 | |
GenBank mRNA | AB209465.1 | - | 12 - 13 | 2129 | 81 |
AK096881.1 | - | 15 - 16 | 1882 | 81 | |
AK290145.1 | - | 15 - 16 | 1816 | 81 | |
AK301832.1 | - | 15 - 16 | 1757 | 81 | |
AK303849.1 | - | 16 - 17 | 1772 | 81 | |
BC001048.2 | - | 15 - 16 | 1895 | 81 | |
BC006190.2 | - | 15 - 16 | 1709 | 81 | |
BC009852.2 | - | 13 - 14 | 1155 | 81 | |
BC015607.2 | - | 15 - 16 | 1862 | 81 | |
X66363.1 | - | 15 - 16 | 1580 | 81 |