The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005264205.4 | XP_005264262.1 | 6 - 7 | 1139 | 71 |
XM_017003586.1 | XP_016859075.1 | 5 - 6 | 1074 | 71 | |
GenBank mRNA | AK301402.1 | - | 5 - 6 | 744 | 71 |
AK301531.1 | - | 6 - 7 | 506 | 71 | |
KU178059.1 | - | 4 - 5 | 638 | 71 |