Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DGKB
Entrez Gene ID
1607
Gene Name
diacylglycerol kinase beta
Gene Aliases
DAGK2, DGK, DGK-BETA
Chromosome Location
Chr.7: 14145049 - 14974777 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004080.2 NP_004071.1 11 - 12 1224 77
NM_145695.2 NP_663733.1 11 - 12 1224 77
XM_005249628.2 XP_005249685.1 12 - 13 1752 77
XM_005249629.2 XP_005249686.1 12 - 13 1752 77
XM_005249630.2 XP_005249687.1 11 - 12 1265 77
XM_005249631.3 XP_005249688.1 11 - 12 1516 77
XM_011515153.2 XP_011513455.1 12 - 13 1590 77
XM_011515154.2 XP_011513456.1 12 - 13 1306 77
XM_011515156.2 XP_011513458.1 11 - 12 1265 77
XM_011515157.2 XP_011513459.1 12 - 13 1305 77
XM_011515158.2 XP_011513460.1 11 - 12 1441 77
XM_017011783.1 XP_016867272.1 12 - 13 1646 77
XM_017011784.1 XP_016867273.1 12 - 13 1306 77
XM_017011785.1 XP_016867274.1 11 - 12 1537 77
XM_017011786.1 XP_016867275.1 11 - 12 1520 77
XM_017011787.1 XP_016867276.1 11 - 12 1270 77
XM_017011788.1 XP_016867277.1 12 - 13 1589 77
XM_017011789.1 XP_016867278.1 12 - 13 1306 77
XM_017011790.1 XP_016867279.1 12 - 13 1589 77
XM_017011791.1 XP_016867280.1 12 - 13 1588 77
XM_017011792.1 XP_016867281.1 12 - 13 1588 77
GenBank mRNA AB018261.1 - 8 - 9 853 77
AK304719.1 - 11 - 12 1516 77
BC105005.1 - 11 - 12 1104 77
BC143632.1 - 11 - 12 1104 77
BC143633.1 - 11 - 12 1104 77

View Full Product Details