The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253852.2 | NP_001240781.1 | 3 - 4 | 678 | 97 |
NM_001253853.2 | NP_001240782.1 | 4 - 5 | 603 | 97 | |
NM_006594.4 | NP_006585.2 | 4 - 5 | 731 | 97 | |
XM_011540523.2 | XP_011538825.1 | 3 - 4 | 584 | 97 | |
XM_011540524.2 | XP_011538826.1 | 2 - 3 | 418 | 97 | |
XM_011540528.2 | XP_011538830.1 | 2 - 3 | 3167 | 97 | |
XM_017000088.1 | XP_016855577.1 | 3 - 4 | 2415 | 97 | |
XM_017000089.1 | XP_016855578.1 | 3 - 4 | 526 | 97 | |
XM_017000092.1 | XP_016855581.1 | 3 - 4 | 526 | 97 | |
XM_017000093.1 | XP_016855582.1 | 3 - 4 | 526 | 97 | |
GenBank mRNA | AB209817.1 | - | 3 - 4 | 615 | 97 |
AK300204.1 | - | 4 - 5 | 442 | 97 | |
AK314641.1 | - | 4 - 5 | 635 | 97 | |
BC014146.2 | - | 3 - 4 | 536 | 97 |