The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032792.3 | NP_116181.1 | 1 - 2 | 302 | 78 |
GenBank mRNA | BC024738.1 | - | 1 - 2 | 297 | 78 |
DA649936.1 | - | 1 - 2 | 302 | 78 |