The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020777.2 | NP_065828.2 | 19 - 20 | 2612 | 77 |
XM_005247987.4 | XP_005248044.2 | 19 - 20 | 2652 | 77 | |
XM_011513514.2 | XP_011511816.1 | 19 - 20 | 2652 | 77 | |
XM_011513515.2 | XP_011511817.1 | 19 - 20 | 2652 | 77 | |
XM_011513516.2 | XP_011511818.1 | 19 - 20 | 2652 | 77 | |
XM_017008481.1 | XP_016863970.1 | 19 - 20 | 2652 | 77 | |
GenBank mRNA | AB037750.1 | - | 16 - 17 | 1856 | 77 |
AK295629.1 | - | 19 - 20 | 2228 | 77 |