Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TRPM6
- Entrez Gene ID
-
140803
- Gene Name
- transient receptor potential cation channel subfamily M member 6
- Gene Aliases
-
CHAK2, HMGX, HOMG, HOMG1, HSH
- Chromosome Location
-
Chr.9: 74722495 - 74888094 on Build GRCh38
View Full Product Details