Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRPM6
Entrez Gene ID
140803
Gene Name
transient receptor potential cation channel subfamily M member 6
Gene Aliases
CHAK2, HMGX, HOMG, HOMG1, HSH
Chromosome Location
Chr.9: 74722495 - 74888094 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177310.1 NP_001170781.1 6 - 7 725 95
NM_001177311.1 NP_001170782.1 6 - 7 752 95
NM_017662.4 NP_060132.3 6 - 7 906 95
XM_011518244.1 XP_011516546.1 6 - 7 906 95
XM_011518245.1 XP_011516547.1 6 - 7 906 95
XM_011518246.1 XP_011516548.1 6 - 7 906 95
XM_011518247.1 XP_011516549.1 6 - 7 906 95
XM_011518248.1 XP_011516550.1 6 - 7 906 95
XM_011518249.1 XP_011516551.1 6 - 7 906 95
XM_011518250.1 XP_011516552.1 6 - 7 906 95
XM_011518252.2 XP_011516554.1 6 - 7 906 95
XM_011518255.2 XP_011516557.1 6 - 7 906 95
XM_017014287.1 XP_016869776.1 6 - 7 906 95
XM_017014288.1 XP_016869777.1 6 - 7 906 95
XM_017014289.1 XP_016869778.1 6 - 7 906 95
GenBank mRNA AF350881.2 - 6 - 7 906 95
AF448232.1 - 6 - 7 668 95
AK057748.1 - 6 - 7 906 95
AY333282.1 - 6 - 7 731 95
AY333283.1 - 6 - 7 725 95
AY333284.1 - 6 - 7 752 95
AY333285.1 - 6 - 7 731 95
AY333286.1 - 6 - 7 731 95
AY333287.1 - 6 - 7 731 95
AY333288.1 - 6 - 7 731 95

View Full Product Details