The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177310.1 | NP_001170781.1 | 25 - 26 | 3593 | 132 |
NM_001177311.1 | NP_001170782.1 | 25 - 26 | 3620 | 132 | |
NM_017662.4 | NP_060132.3 | 25 - 26 | 3774 | 132 | |
XM_011518244.1 | XP_011516546.1 | 25 - 26 | 3774 | 132 | |
XM_011518245.1 | XP_011516547.1 | 24 - 25 | 3681 | 132 | |
XM_011518246.1 | XP_011516548.1 | 25 - 26 | 3774 | 132 | |
XM_011518247.1 | XP_011516549.1 | 24 - 25 | 3645 | 132 | |
XM_011518248.1 | XP_011516550.1 | 24 - 25 | 3633 | 132 | |
XM_011518249.1 | XP_011516551.1 | 23 - 24 | 3540 | 132 | |
XM_011518250.1 | XP_011516552.1 | 23 - 24 | 3498 | 132 | |
XM_011518251.2 | XP_011516553.1 | 20 - 21 | 3025 | 132 | |
XM_011518252.2 | XP_011516554.1 | 25 - 26 | 3774 | 132 | |
XM_017014287.1 | XP_016869776.1 | 22 - 23 | 3411 | 132 | |
XM_017014288.1 | XP_016869777.1 | 21 - 22 | 3264 | 132 | |
XM_017014289.1 | XP_016869778.1 | 25 - 26 | 3774 | 132 | |
GenBank mRNA | AF350881.2 | - | 25 - 26 | 3774 | 132 |
AF448232.1 | - | 25 - 26 | 3536 | 132 | |
AK000094.1 | - | 2 - 3 | 500 | 132 | |
AK026281.1 | - | 19 - 20 | 2841 | 132 | |
AK225207.1 | - | 2 - 3 | 500 | 132 | |
AY333282.1 | - | 25 - 26 | 3599 | 132 | |
AY333283.1 | - | 25 - 26 | 3593 | 132 | |
AY333284.1 | - | 25 - 26 | 3620 | 132 | |
AY333285.1 | - | 25 - 26 | 3599 | 132 |