The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177310.1 | NP_001170781.1 | 37 - 38 | 5877 | 83 |
NM_001177311.1 | NP_001170782.1 | 37 - 38 | 5904 | 83 | |
NM_017662.4 | NP_060132.3 | 37 - 38 | 6058 | 83 | |
XM_011518244.1 | XP_011516546.1 | 37 - 38 | 6055 | 83 | |
XM_011518245.1 | XP_011516547.1 | 36 - 37 | 5965 | 83 | |
XM_011518246.1 | XP_011516548.1 | 38 - 39 | 5935 | 83 | |
XM_011518247.1 | XP_011516549.1 | 36 - 37 | 5929 | 83 | |
XM_011518248.1 | XP_011516550.1 | 36 - 37 | 5917 | 83 | |
XM_011518249.1 | XP_011516551.1 | 35 - 36 | 5824 | 83 | |
XM_011518250.1 | XP_011516552.1 | 35 - 36 | 5782 | 83 | |
XM_011518251.2 | XP_011516553.1 | 32 - 33 | 5309 | 83 | |
XM_017014287.1 | XP_016869776.1 | 34 - 35 | 5695 | 83 | |
XM_017014288.1 | XP_016869777.1 | 33 - 34 | 5548 | 83 | |
GenBank mRNA | AF350881.2 | - | 37 - 38 | 6058 | 83 |
AF448232.1 | - | 37 - 38 | 5820 | 83 | |
AK000094.1 | - | 14 - 15 | 2784 | 83 | |
AK225207.1 | - | 14 - 15 | 2784 | 83 | |
AY333282.1 | - | 37 - 38 | 5883 | 83 | |
AY333283.1 | - | 37 - 38 | 5877 | 83 | |
AY333284.1 | - | 37 - 38 | 5904 | 83 | |
AY333286.1 | - | 24 - 25 | 2736 | 83 | |
AY333287.1 | - | 21 - 22 | 2382 | 83 | |
AY333288.1 | - | 11 - 12 | 1524 | 83 |