The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037872.2 | NP_001032961.1 | 11 - 12 | 2073 | 114 |
NM_001321454.1 | NP_001308383.1 | 12 - 13 | 2184 | 114 | |
NM_001321455.1 | NP_001308384.1 | 12 - 13 | 2230 | 114 | |
NM_001321458.1 | NP_001308387.1 | 12 - 13 | 2200 | 114 | |
NM_001321459.1 | NP_001308388.1 | 12 - 13 | 2276 | 114 | |
NM_001321460.1 | NP_001308389.1 | 12 - 13 | 2273 | 114 | |
NM_016316.3 | NP_057400.1 | 11 - 12 | 2076 | 114 | |
NR_135649.1 | 12 - 13 | 2273 | 114 | ||
NR_135650.1 | 12 - 13 | 2230 | 114 | ||
NR_135651.1 | 11 - 12 | 2073 | 114 | ||
NR_135652.1 | 11 - 12 | 2076 | 114 | ||
NR_135653.1 | 11 - 12 | 2076 | 114 | ||
XM_011511341.2 | XP_011509643.1 | 5 - 6 | 693 | 114 | |
XM_017004309.1 | XP_016859798.1 | 12 - 13 | 2597 | 114 | |
XM_017004310.1 | XP_016859799.1 | 12 - 13 | 1963 | 114 | |
XM_017004311.1 | XP_016859800.1 | 12 - 13 | 1963 | 114 | |
XM_017004312.1 | XP_016859801.1 | 12 - 13 | 2176 | 114 | |
XM_017004313.1 | XP_016859802.1 | 11 - 12 | 3802 | 114 | |
XM_017004314.1 | XP_016859803.1 | 8 - 9 | 1453 | 114 | |
XM_017004315.1 | XP_016859804.1 | 8 - 9 | 1437 | 114 | |
GenBank mRNA | AB047646.1 | - | 11 - 12 | 1998 | 114 |
AF151538.1 | - | 11 - 12 | 2029 | 114 | |
AF206019.1 | - | 11 - 12 | 2038 | 114 | |
AF357886.1 | - | 11 - 12 | 1994 | 114 | |
AJ131720.1 | - | 3 - 4 | 395 | 114 | |
AK025176.1 | - | 3 - 4 | 355 | 114 | |
AK307719.1 | - | 11 - 12 | 1987 | 114 | |
BC037734.1 | - | 12 - 13 | 2211 | 114 | |
BC130411.1 | - | 10 - 11 | 1844 | 114 |