Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
REV1
Entrez Gene ID
51455
Gene Name
REV1, DNA directed polymerase
Gene Aliases
AIBP80, REV1L
Chromosome Location
Chr.2: 99400475 - 99490123 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037872.2 NP_001032961.1 4 - 5 604 129
NM_001321454.1 NP_001308383.1 4 - 5 604 129
NM_001321455.1 NP_001308384.1 5 - 6 761 129
NM_001321458.1 NP_001308387.1 4 - 5 604 129
NM_001321459.1 NP_001308388.1 4 - 5 604 129
NM_001321460.1 NP_001308389.1 4 - 5 604 129
NM_016316.3 NP_057400.1 4 - 5 604 129
NR_135649.1 4 - 5 604 129
NR_135650.1 5 - 6 761 129
NR_135651.1 4 - 5 604 129
NR_135652.1 4 - 5 604 129
NR_135653.1 4 - 5 604 129
XM_017004309.1 XP_016859798.1 4 - 5 1017 129
XM_017004310.1 XP_016859799.1 4 - 5 383 129
XM_017004311.1 XP_016859800.1 4 - 5 383 129
XM_017004312.1 XP_016859801.1 4 - 5 599 129
XM_017004313.1 XP_016859802.1 4 - 5 2333 129
GenBank mRNA AB047646.1 - 4 - 5 529 129
AF151538.1 - 4 - 5 557 129
AF206019.1 - 4 - 5 566 129
AF357886.1 - 4 - 5 522 129
AK002087.1 - 2 - 3 466 129
AK307719.1 - 4 - 5 515 129
BC037734.1 - 5 - 6 742 129
BC130411.1 - 3 - 4 375 129
DR000409.1 - 4 - 5 495 129

View Full Product Details