The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032663.4 | NP_116052.2 | 1 - 2 | 184 | 86 |
XM_005253962.3 | XP_005254019.1 | 1 - 2 | 170 | 86 | |
XM_005253965.4 | XP_005254022.1 | 1 - 2 | 178 | 86 | |
XM_017020048.1 | XP_016875537.1 | 1 - 2 | 179 | 86 | |
XM_017020049.1 | XP_016875538.1 | 1 - 2 | 170 | 86 | |
XM_017020053.1 | XP_016875542.1 | 1 - 2 | 94 | 86 | |
GenBank mRNA | AJ586136.1 | - | 1 - 2 | 75 | 86 |
BC004868.2 | - | 1 - 2 | 174 | 86 |