Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WWC2
Entrez Gene ID
80014
Gene Name
WW and C2 domain containing 2
Gene Aliases
BOMB
Chromosome Location
Chr.4: 183098677 - 183320777 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024949.5 NP_079225.5 12 - 13 2213 111
XM_011532269.2 XP_011530571.1 12 - 13 2846 111
GenBank mRNA AK126057.1 - 11 - 12 1878 111
AK127061.1 - 5 - 6 1125 111
BC053873.1 - 3 - 4 642 111
BC107683.1 - 3 - 4 810 111
BX647378.1 - 10 - 11 1804 111

View Full Product Details