The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003074.3 | NP_003065.3 | 22 - 23 | 2499 | 71 |
XM_011534034.1 | XP_011532336.1 | 22 - 23 | 2230 | 71 | |
GenBank mRNA | AK302541.1 | - | 23 - 24 | 2274 | 71 |
BC021862.1 | - | 22 - 23 | 2436 | 71 | |
BC032564.1 | - | 22 - 23 | 2483 | 71 | |
BC039843.1 | - | 22 - 23 | 2403 | 71 | |
BC040242.1 | - | 7 - 8 | 931 | 71 | |
BC050564.1 | - | 22 - 23 | 2428 | 71 | |
BC065253.1 | - | 22 - 23 | 2468 | 71 | |
BC113465.1 | - | 22 - 23 | 2447 | 71 | |
BC117213.1 | - | 22 - 23 | 2447 | 71 | |
U66615.1 | - | 22 - 23 | 2432 | 71 |