The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003074.3 | NP_003065.3 | 24 - 25 | 2766 | 68 |
XM_011534034.1 | XP_011532336.1 | 24 - 25 | 2497 | 68 | |
GenBank mRNA | AK302412.1 | - | 11 - 12 | 1210 | 68 |
AK302541.1 | - | 25 - 26 | 2541 | 68 | |
BC040242.1 | - | 9 - 10 | 1198 | 68 | |
BC050564.1 | - | 24 - 25 | 2695 | 68 | |
BC113465.1 | - | 24 - 25 | 2714 | 68 | |
BC117213.1 | - | 24 - 25 | 2714 | 68 | |
U66615.1 | - | 24 - 25 | 2699 | 68 |