The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015691.3 | NP_056506.2 | 21 - 22 | 3282 | 110 |
GenBank mRNA | AB033106.1 | - | 19 - 20 | 2905 | 110 |
AK024683.1 | - | 12 - 13 | 2092 | 110 | |
AK292961.1 | - | 20 - 21 | 3164 | 110 | |
AL353937.1 | - | 18 - 19 | 2773 | 110 | |
BC003527.1 | - | 3 - 4 | 459 | 110 | |
BC032447.1 | - | 18 - 19 | 2817 | 110 | |
KC987947.1 | - | 21 - 22 | 3459 | 110 |