Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WWC3
Entrez Gene ID
55841
Gene Name
WWC family member 3
Gene Aliases
BM042
Chromosome Location
Chr.X: 10015755 - 10144478 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015691.3 NP_056506.2 22 - 23 3404 64
GenBank mRNA AB033106.1 - 20 - 21 3027 64
AK024683.1 - 13 - 14 2214 64
AK091936.1 - 13 - 14 2081 64
AK292961.1 - 21 - 22 3286 64
AL353937.1 - 19 - 20 2895 64
BC003527.1 - 4 - 5 581 64
BC032447.1 - 19 - 20 2939 64
KC987947.1 - 22 - 23 3581 64

View Full Product Details