Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCSK1
Entrez Gene ID
5122
Gene Name
proprotein convertase subtilisin/kexin type 1
Gene Aliases
BMIQ12, NEC1, PC1, PC3, SPC3
Chromosome Location
Chr.5: 96390336 - 96433281 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000439.4 NP_000430.3 3 - 4 634 58
NM_001177875.1 NP_001171346.1 3 - 4 292 58
GenBank mRNA AB208874.1 - 3 - 4 310 58
AK303888.1 - 3 - 4 292 58
BC130295.1 - 3 - 4 516 58
BC136486.1 - 3 - 4 516 58
M90753.1 - 3 - 4 604 58
X64810.1 - 3 - 4 584 58

View Full Product Details