Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZNF84
Entrez Gene ID
7637
Gene Name
zinc finger protein 84
Gene Aliases
HPF2
Chromosome Location
Chr.12: 133037286 - 133063304 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127372.2 NP_001120844.1 3 - 4 600 121
NM_001289971.1 NP_001276900.1 3 - 4 591 121
NM_001289972.1 NP_001276901.1 3 - 4 719 121
NM_003428.5 NP_003419.3 3 - 4 584 121
XM_005266185.1 XP_005266242.1 3 - 4 754 121
XM_005266186.1 XP_005266243.1 4 - 5 490 121
XM_011534832.1 XP_011533134.1 3 - 4 534 121
GenBank mRNA AK314232.1 - 3 - 4 362 121
BC036656.1 - 3 - 4 706 121
BC048350.1 - 2 - 3 168 121
BC062552.1 - 3 - 4 360 121
DA663778.1 - 3 - 4 492 121
DA739478.1 - 3 - 4 478 121
X60156.2 - 2 - 3 330 121

View Full Product Details