The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135662.1 | NP_001129134.1 | 4 - 5 | 616 | 63 |
NM_001135663.1 | NP_001129135.1 | 2 - 3 | 831 | 63 | |
NM_001135664.1 | NP_001129136.1 | 3 - 4 | 463 | 63 | |
NM_003929.2 | NP_003920.1 | 4 - 5 | 717 | 63 | |
XM_005245569.1 | XP_005245626.1 | 4 - 5 | 615 | 63 | |
XM_005245570.1 | XP_005245627.1 | 4 - 5 | 668 | 63 | |
XM_005245571.1 | XP_005245628.1 | 4 - 5 | 593 | 63 | |
XM_006711605.3 | XP_006711668.1 | 3 - 4 | 359 | 63 | |
XM_006711606.2 | XP_006711669.1 | 3 - 4 | 323 | 63 | |
XM_017002748.1 | XP_016858237.1 | 3 - 4 | 618 | 63 | |
XM_017002749.1 | XP_016858238.1 | 3 - 4 | 596 | 63 | |
XM_017002750.1 | XP_016858239.1 | 3 - 4 | 528 | 63 | |
GenBank mRNA | AK223514.1 | - | 3 - 4 | 493 | 63 |
AK303879.1 | - | 2 - 3 | 582 | 63 | |
AK308359.1 | - | 3 - 4 | 432 | 63 | |
AK313000.1 | - | 4 - 5 | 605 | 63 | |
BC002585.2 | - | 4 - 5 | 550 | 63 | |
BT019540.1 | - | 3 - 4 | 377 | 63 | |
BX647960.1 | - | 4 - 5 | 682 | 63 | |
CR542010.1 | - | 3 - 4 | 377 | 63 | |
D84488.1 | - | 3 - 4 | 417 | 63 | |
DA377295.1 | - | 3 - 4 | 418 | 63 |