Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPATA7
Entrez Gene ID
55812
Gene Name
spermatogenesis associated 7
Gene Aliases
HEL-S-296, HSD-3.1, HSD3, LCA3
Chromosome Location
Chr.14: 88385638 - 88470350 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018418.4 NP_060888.2 3 - 4 367 116
XM_005267851.1 XP_005267908.1 3 - 4 338 116
XM_006720204.1 XP_006720267.1 3 - 4 338 116
XM_006720205.1 XP_006720268.1 3 - 4 338 116
XM_011536951.1 XP_011535253.1 3 - 4 532 116
XM_017021452.1 XP_016876941.1 3 - 4 532 116
GenBank mRNA AF144487.3 - 3 - 4 347 116
AF144488.2 - 1 - 2 72 116
AK055864.1 - 3 - 4 289 116
AK225746.1 - 4 - 5 338 116
AK293007.1 - 4 - 5 320 116
CR936777.1 - 3 - 4 313 116
EU668353.1 - 3 - 4 343 116

View Full Product Details