The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040428.3 | NP_001035518.1 | 3 - 4 | 320 | 134 |
NM_018418.4 | NP_060888.2 | 4 - 5 | 416 | 134 | |
XM_005267851.1 | XP_005267908.1 | 4 - 5 | 387 | 134 | |
XM_005267852.1 | XP_005267909.1 | 3 - 4 | 316 | 134 | |
XM_006720204.1 | XP_006720267.1 | 4 - 5 | 387 | 134 | |
XM_006720205.1 | XP_006720268.1 | 4 - 5 | 387 | 134 | |
XM_011536951.1 | XP_011535253.1 | 4 - 5 | 581 | 134 | |
XM_011536952.1 | XP_011535254.1 | 2 - 3 | 199 | 134 | |
XM_011536953.1 | XP_011535255.1 | 3 - 4 | 520 | 134 | |
XM_017021452.1 | XP_016876941.1 | 4 - 5 | 581 | 134 | |
XM_017021457.1 | XP_016876946.1 | 3 - 4 | 520 | 134 | |
GenBank mRNA | AF144487.3 | - | 4 - 5 | 396 | 134 |
AF144488.2 | - | 2 - 3 | 121 | 134 | |
AK055864.1 | - | 4 - 5 | 338 | 134 | |
AK225746.1 | - | 5 - 6 | 387 | 134 | |
AK290631.1 | - | 3 - 4 | 291 | 134 | |
AK293007.1 | - | 5 - 6 | 369 | 134 | |
AL136604.1 | - | 3 - 4 | 300 | 134 | |
BC090875.1 | - | 4 - 5 | 703 | 134 | |
CR533563.1 | - | 3 - 4 | 145 | 134 | |
CR936777.1 | - | 4 - 5 | 362 | 134 | |
EU668353.1 | - | 4 - 5 | 392 | 134 |