The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271885.1 | NP_001258814.1 | 9 - 10 | 861 | 62 |
NM_001271886.1 | NP_001258815.1 | 9 - 10 | 1122 | 62 | |
NM_024666.4 | NP_078942.3 | 9 - 10 | 977 | 62 | |
XM_011522020.1 | XP_011520322.1 | 9 - 10 | 1227 | 62 | |
GenBank mRNA | AK021568.1 | - | 9 - 10 | 881 | 62 |
AK294402.1 | - | 9 - 10 | 858 | 62 | |
AL136715.1 | - | 9 - 10 | 887 | 62 | |
AL832256.1 | - | 6 - 7 | 558 | 62 | |
BC001975.1 | - | 6 - 7 | 498 | 62 | |
BC047026.1 | - | 9 - 10 | 900 | 62 | |
BC058886.1 | - | 9 - 10 | 899 | 62 | |
CR533540.1 | - | 9 - 10 | 872 | 62 |