Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRDM8
Entrez Gene ID
56978
Gene Name
PR domain 8
Gene Aliases
EPM10, PFM5
Chromosome Location
Chr.4: 80183418 - 80204329 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020226.3 NP_064611.3 4 - 5 589 116
XM_011532133.2 XP_011530435.1 3 - 4 880 116
XM_011532140.2 XP_011530442.1 3 - 4 448 116
XM_017008468.1 XP_016863957.1 3 - 4 331 116
XM_017008470.1 XP_016863959.1 3 - 4 331 116
GenBank mRNA AF275815.1 - 4 - 5 591 116

View Full Product Details