Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACTR3
Entrez Gene ID
10096
Gene Name
ARP3 actin related protein 3 homolog
Gene Aliases
ARP3
Chromosome Location
Chr.2: 113889934 - 113961554 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277140.1 NP_001264069.1 2 - 3 235 70
NM_005721.4 NP_005712.1 2 - 3 444 70
GenBank mRNA AF006083.1 - 2 - 3 270 70
AF127773.1 - 2 - 3 313 70
AK302152.1 - 2 - 3 189 70
AK302359.1 - 2 - 3 223 70
AK303988.1 - 2 - 3 281 70
AK312659.1 - 2 - 3 244 70
AK316213.1 - 2 - 3 266 70
AK316396.1 - 2 - 3 235 70
BC044590.1 - 2 - 3 418 70
DB453995.1 - 2 - 3 444 70

View Full Product Details