The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004272.4 | NP_004263.1 | 5 - 6 | 1593 | 71 |
GenBank mRNA | AF093262.1 | - | 4 - 5 | 531 | 71 |
AK223471.1 | - | 5 - 6 | 855 | 71 | |
AK312481.1 | - | 5 - 6 | 858 | 71 | |
BC015502.1 | - | 5 - 6 | 808 | 71 | |
BC047438.1 | - | 5 - 6 | 1593 | 71 | |
BT009846.1 | - | 4 - 5 | 531 | 71 | |
Y17829.1 | - | 5 - 6 | 606 | 71 |