Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HOMER1
Entrez Gene ID
9456
Gene Name
homer scaffolding protein 1
Gene Aliases
HOMER, HOMER1A, HOMER1B, HOMER1C, SYN47, Ves-1
Chromosome Location
Chr.5: 79373824 - 79514568 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277077.1 NP_001264006.1 5 - 6 1551 139
NM_004272.4 NP_004263.1 8 - 9 1941 139
GenBank mRNA AF093262.1 - 7 - 8 879 139
AK223471.1 - 8 - 9 1203 139
AK312481.1 - 8 - 9 1206 139
AY189939.1 - 5 - 6 509 139
AY189940.1 - 4 - 5 377 139
BC015502.1 - 8 - 9 1156 139
BT009846.1 - 7 - 8 879 139
Y17829.1 - 8 - 9 954 139

View Full Product Details