The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033057.1 | NP_001028229.1 | 17 - 18 | 3519 | 82 |
NM_015520.1 | NP_056335.1 | 18 - 19 | 3603 | 82 | |
XM_005265563.1 | XP_005265620.1 | 18 - 19 | 3611 | 82 | |
XM_005265568.1 | XP_005265625.1 | 17 - 18 | 3569 | 82 | |
XM_017007497.1 | XP_016862986.1 | 18 - 19 | 3608 | 82 | |
XM_017007498.1 | XP_016862987.1 | 18 - 19 | 3605 | 82 | |
XM_017007500.1 | XP_016862989.1 | 18 - 19 | 3959 | 82 | |
XM_017007503.1 | XP_016862992.1 | 18 - 19 | 3611 | 82 | |
XM_017007504.1 | XP_016862993.1 | 18 - 19 | 3608 | 82 | |
XM_017007505.1 | XP_016862994.1 | 18 - 19 | 3608 | 82 | |
GenBank mRNA | AF401654.1 | - | 18 - 19 | 3077 | 82 |
AF401656.1 | - | 17 - 18 | 2993 | 82 |