Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAGI1
Entrez Gene ID
9223
Gene Name
membrane associated guanylate kinase, WW and PDZ domain containing 1
Gene Aliases
AIP-3, AIP3, BAIAP1, BAP-1, BAP1, MAGI-1, Magi1d, TNRC19, WWP3
Chromosome Location
Chr.3: 65353525 - 66038836 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033057.1 NP_001028229.1 22 - 23 4166 113
XM_005265563.1 XP_005265620.1 23 - 24 4258 113
XM_005265564.1 XP_005265621.1 23 - 24 4255 113
XM_005265565.1 XP_005265622.1 23 - 24 4225 113
XM_005265566.1 XP_005265623.1 23 - 24 4219 113
XM_005265568.1 XP_005265625.1 22 - 23 4216 113
XM_005265570.1 XP_005265627.1 22 - 23 4057 113
XM_005265571.1 XP_005265628.1 21 - 22 3967 113
XM_006713407.1 XP_006713470.1 23 - 24 4261 113
XM_006713408.1 XP_006713471.1 23 - 24 4258 113
XM_006713409.1 XP_006713472.1 23 - 24 4258 113
XM_006713410.1 XP_006713473.1 22 - 23 4219 113
XM_006713411.1 XP_006713474.1 22 - 23 4177 113
XM_006713412.1 XP_006713475.1 22 - 23 4171 113
XM_011534236.1 XP_011532538.1 22 - 23 4222 113
XM_011534240.2 XP_011532542.1 11 - 12 5617 113
XM_011534241.2 XP_011532543.1 10 - 11 5531 113
XM_017007497.1 XP_016862986.1 23 - 24 4255 113
XM_017007498.1 XP_016862987.1 23 - 24 4252 113
XM_017007499.1 XP_016862988.1 23 - 24 4616 113
XM_017007500.1 XP_016862989.1 23 - 24 4606 113
XM_017007501.1 XP_016862990.1 22 - 23 4138 113
XM_017007507.1 XP_016862996.1 21 - 22 3299 113
XM_017007508.1 XP_016862997.1 20 - 21 3131 113
GenBank mRNA AF401656.1 - 22 - 23 3640 113
KC170987.1 - 20 - 21 3145 113

View Full Product Details