The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001520.2 | NP_001001520.1 | 4 - 5 | 640 | 62 |
NM_032631.3 | NP_116020.1 | 4 - 5 | 640 | 62 | |
XM_011528375.2 | XP_011526677.1 | 4 - 5 | 1000 | 62 | |
XM_011528376.2 | XP_011526678.1 | 4 - 5 | 1000 | 62 | |
XM_011528377.2 | XP_011526679.1 | 4 - 5 | 1000 | 62 | |
GenBank mRNA | AB208892.1 | - | 2 - 3 | 339 | 62 |
AF294267.1 | - | 4 - 5 | 547 | 62 | |
AL520441.3 | - | 4 - 5 | 585 | 62 | |
AY358600.1 | - | 4 - 5 | 549 | 62 | |
BC000755.2 | - | 4 - 5 | 530 | 62 | |
BC009449.1 | - | 1 - 2 | 179 | 62 | |
BI905780.1 | - | 4 - 5 | 578 | 62 |