Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARHGEF7
Entrez Gene ID
8874
Gene Name
Rho guanine nucleotide exchange factor 7
Gene Aliases
BETA-PIX, COOL-1, COOL1, Nbla10314, P50, P50BP, P85, P85COOL1, P85SPR, PAK3, PIXB
Chromosome Location
Chr.13: 111114619 - 111305734 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113511.2 NP_001106983.1 14 - 15 2476 67
NM_001113512.2 NP_001106984.1 12 - 13 2326 67
NM_001113513.1 NP_001106985.1 12 - 13 1418 67
NM_001320851.1 NP_001307780.1 11 - 12 1199 67
NM_001320852.1 NP_001307781.1 13 - 14 2413 67
NM_001320853.1 NP_001307782.1 12 - 13 1324 67
NM_001320854.1 NP_001307783.1 8 - 9 938 67
NM_003899.3 NP_003890.1 12 - 13 1531 67
NM_145735.3 NP_663788.1 13 - 14 2413 67
XM_006719956.3 XP_006720019.1 13 - 14 1541 67
XM_017020814.1 XP_016876303.1 13 - 14 1541 67
XM_017020815.1 XP_016876304.1 11 - 12 1534 67
XM_017020816.1 XP_016876305.1 12 - 13 1309 67
XM_017020817.1 XP_016876306.1 11 - 12 1534 67
XM_017020818.1 XP_016876307.1 13 - 14 1409 67
XM_017020819.1 XP_016876308.1 12 - 13 1418 67
XM_017020820.1 XP_016876309.1 12 - 13 1541 67
XM_017020821.1 XP_016876310.1 12 - 13 1236 67
XM_017020822.1 XP_016876311.1 11 - 12 1232 67
XM_017020823.1 XP_016876312.1 11 - 12 2009 67
XM_017020824.1 XP_016876313.1 13 - 14 1409 67
XM_017020825.1 XP_016876314.1 11 - 12 1160 67
XM_017020826.1 XP_016876315.1 13 - 14 1409 67
XM_017020827.1 XP_016876316.1 11 - 12 1233 67
XM_017020828.1 XP_016876317.1 9 - 10 857 67
XM_017020829.1 XP_016876318.1 9 - 10 857 67
GenBank mRNA AB075521.1 - 3 - 4 272 67
AB177848.1 - 13 - 14 2414 67
AB177849.1 - 10 - 11 1137 67
AK295472.1 - 8 - 9 863 67
AK300272.1 - 12 - 13 1311 67
AL834228.1 - 13 - 14 1597 67
BC033905.1 - 12 - 13 1523 67
BC050521.1 - 12 - 13 1390 67
BC060776.1 - 14 - 15 1818 67
D63476.1 - 12 - 13 1507 67

View Full Product Details