The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113513.1 | NP_001106985.1 | 19 - 20 | 2199 | 101 |
NM_001320851.1 | NP_001307780.1 | 18 - 19 | 1980 | 101 | |
NM_001320852.1 | NP_001307781.1 | 20 - 21 | 3194 | 101 | |
NM_001320853.1 | NP_001307782.1 | 19 - 20 | 2105 | 101 | |
NM_003899.3 | NP_003890.1 | 19 - 20 | 2312 | 101 | |
XM_006719956.3 | XP_006720019.1 | 20 - 21 | 2358 | 101 | |
XM_017020815.1 | XP_016876304.1 | 18 - 19 | 2351 | 101 | |
XM_017020816.1 | XP_016876305.1 | 19 - 20 | 2126 | 101 | |
XM_017020818.1 | XP_016876307.1 | 20 - 21 | 2226 | 101 | |
XM_017020819.1 | XP_016876308.1 | 19 - 20 | 2235 | 101 | |
XM_017020820.1 | XP_016876309.1 | 19 - 20 | 2358 | 101 | |
XM_017020821.1 | XP_016876310.1 | 19 - 20 | 2053 | 101 | |
XM_017020822.1 | XP_016876311.1 | 18 - 19 | 2049 | 101 | |
XM_017020823.1 | XP_016876312.1 | 18 - 19 | 2826 | 101 | |
XM_017020824.1 | XP_016876313.1 | 20 - 21 | 2190 | 101 | |
XM_017020825.1 | XP_016876314.1 | 18 - 19 | 1941 | 101 | |
XM_017020828.1 | XP_016876317.1 | 16 - 17 | 1674 | 101 | |
GenBank mRNA | AB177849.1 | - | 17 - 18 | 1918 | 101 |
AK300272.1 | - | 19 - 20 | 2092 | 101 | |
BC050521.1 | - | 19 - 20 | 2171 | 101 | |
D63476.1 | - | 19 - 20 | 2288 | 101 |