Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARHGEF7
Entrez Gene ID
8874
Gene Name
Rho guanine nucleotide exchange factor 7
Gene Aliases
BETA-PIX, COOL-1, COOL1, Nbla10314, P50, P50BP, P85, P85COOL1, P85SPR, PAK3, PIXB
Chromosome Location
Chr.13: 111114619 - 111305734 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113511.2 NP_001106983.1 6 - 7 1641 55
NM_001113512.2 NP_001106984.1 4 - 5 1491 55
NM_001113513.1 NP_001106985.1 4 - 5 583 55
NM_001320851.1 NP_001307780.1 3 - 4 364 55
NM_001320852.1 NP_001307781.1 5 - 6 1578 55
NM_001320853.1 NP_001307782.1 4 - 5 489 55
NM_003899.3 NP_003890.1 4 - 5 696 55
NM_145735.3 NP_663788.1 5 - 6 1578 55
XM_006719956.3 XP_006720019.1 5 - 6 706 55
XM_011521133.2 XP_011519435.1 5 - 6 706 55
XM_017020814.1 XP_016876303.1 5 - 6 706 55
XM_017020815.1 XP_016876304.1 3 - 4 699 55
XM_017020816.1 XP_016876305.1 4 - 5 474 55
XM_017020817.1 XP_016876306.1 3 - 4 699 55
XM_017020818.1 XP_016876307.1 5 - 6 574 55
XM_017020819.1 XP_016876308.1 4 - 5 583 55
XM_017020820.1 XP_016876309.1 4 - 5 706 55
XM_017020821.1 XP_016876310.1 4 - 5 401 55
XM_017020822.1 XP_016876311.1 3 - 4 397 55
XM_017020823.1 XP_016876312.1 3 - 4 1174 55
XM_017020824.1 XP_016876313.1 5 - 6 574 55
XM_017020825.1 XP_016876314.1 3 - 4 325 55
XM_017020826.1 XP_016876315.1 5 - 6 574 55
XM_017020827.1 XP_016876316.1 3 - 4 398 55
GenBank mRNA AB177848.1 - 5 - 6 1579 55
AB177849.1 - 2 - 3 302 55
AJ844629.1 - 1 - 2 26 55
AK300157.1 - 4 - 5 407 55
AK300272.1 - 4 - 5 476 55
AL834228.1 - 5 - 6 762 55
BC033905.1 - 4 - 5 688 55
BC050521.1 - 4 - 5 555 55
BC060776.1 - 6 - 7 983 55
D63476.1 - 4 - 5 672 55
DC318118.1 - 4 - 5 489 55

View Full Product Details