Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYLD
Entrez Gene ID
1540
Gene Name
CYLD lysine 63 deubiquitinase
Gene Aliases
BRSS, CDMT, CYLD1, CYLDI, EAC, MFT, MFT1, SBS, TEM, USPL2
Chromosome Location
Chr.16: 50742026 - 50801935 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042355.1 NP_001035814.1 3 - 4 782 71
NM_001042412.1 NP_001035877.1 3 - 4 806 71
NM_015247.2 NP_056062.1 4 - 5 919 71
XM_005255812.2 XP_005255869.1 4 - 5 879 71
XM_006721149.1 XP_006721212.1 4 - 5 943 71
XM_011522907.2 XP_011521209.1 3 - 4 705 71
XM_011522908.1 XP_011521210.1 3 - 4 797 71
XM_011522911.2 XP_011521213.1 3 - 4 797 71
XM_017022977.1 XP_016878466.1 3 - 4 1077 71
XM_017022978.1 XP_016878467.1 3 - 4 873 71
XM_017022979.1 XP_016878468.1 3 - 4 833 71
XM_017022980.1 XP_016878469.1 3 - 4 866 71
XM_017022981.1 XP_016878470.1 3 - 4 764 71
GenBank mRNA AJ250014.1 - 4 - 5 895 71
AK024212.1 - 1 - 2 240 71
AK024348.1 - 1 - 2 289 71
AK056226.1 - 3 - 4 737 71
AK225202.1 - 4 - 5 899 71
AK292975.1 - 3 - 4 764 71
AK310832.1 - 3 - 4 754 71
BC012342.1 - 3 - 4 780 71

View Full Product Details