The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005263144.2 | XP_005263201.1 | 3 - 4 | 393 | 135 |
XM_005263146.4 | XP_005263203.1 | 2 - 3 | 197 | 135 | |
GenBank mRNA | BC058908.1 | - | 2 - 3 | 185 | 135 |